man

Test

CAS RN 1467057-23-3

CAS RN 1467057-23-3

IUPAC Name:
Synonyms:
Molecular Weight (g/mol):
Molecular Formula:
InChi Key:
SMILES:
Color 
  • (4)
Product Type 
  • (4)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (4)