man

Test

CAS RN 154447-35-5

ONOO

CAS RN 154447-35-5

IUPAC Name: 2-(morpholin-4-yl)-4H-benzo[h]chromen-4-one
Synonyms: 2-(morpholin-4-yl)benzo[h]chromen-4-one
Molecular Weight (g/mol): 281.31
Molecular Formula: C17H15NO3
InChi Key: KKTZALUTXUZPSN-UHFFFAOYSA-N
SMILES: O=C1C=C(OC2=C1C=CC1=CC=CC=C21)N1CCOCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results