missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromophenyl)pyrrolidine, 97%, Thermo Scientific™

255.34 SEK - 2814.44 SEK

Chemical Identifiers

CAS 22090-26-2
Molecular Formula C10H12BrN
Molecular Weight (g/mol) 226.117
InChI Key BVEGBJXZICCEQW-UHFFFAOYSA-N
Synonym 1-4-bromophenyl pyrrolidine, n-4-bromophenyl pyrrolidine, 1-4-bromophenyl-pyrrolidine, acmc-209frt, 4-pyrrolidinobromobenzene, 4-bromophenyl pyrrolidine, 1-4-bromo-phenyl-pyrrolidine, pyrrolidine,1-4-bromophenyl, 4-bromo-1-1-pyrrolidinyl benzeneShow MoreShow Less
PubChem CID 7016457
IUPAC Name 1-(4-bromophenyl)pyrrolidine
SMILES C1CCN(C1)C2=CC=C(C=C2)Br
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10239942
SDS missing translation for 'viewDocuments'
Thermo Scientific Maybridge
CC30110CB
250 mg
255.34 SEK
250mg
Please to purchase this item. Need a web account? Register with us today!
10691265
SDS missing translation for 'viewDocuments'
Thermo Scientific Maybridge
CC30110DA
1 g
365.42 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
10356284
SDS missing translation for 'viewDocuments'
Thermo Scientific Maybridge
CC30110EA
10 g
2814.44 SEK
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(4-Bromophenyl)pyrrolidine, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.