missing translation for 'onlineSavingsMsg'
Learn More
1-[4-(tert-Butyl)phenyl]ethan-1-one, 97%, Thermo Scientific™ Product Code.: 10279103

1-[4-(tert-Butyl)phenyl]ethan-1-one, 97%, Thermo Scientific™

Product Code. 10279103
10 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 10279103

Brand: Thermo Scientific Maybridge BTB09870EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 943-27-1
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
InChI Key UYFJYGWNYQCHOB-UHFFFAOYSA-N
Synonym 4'-tert-butylacetophenone, 1-4-tert-butylphenyl ethanone, 4-tert-butylacetophenone, 1-4-tert-butyl phenyl ethanone, p-tert-butylacetophenone, ethanone, 1-4-1,1-dimethylethyl phenyl, acetophenone, 4'-tert-butyl, 1-4-tert-butyl phenyl ethan-1-one, p-t-butylacetophenone, 1-4-tert-butyl-phenyl-ethanoneShow MoreShow Less
PubChem CID 13669
IUPAC Name 1-(4-tert-butylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)C(C)(C)C

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 176.26
Chemical Name or Material 1-[4-(tert-Butyl)phenyl]ethan-1-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-[4-(tert-Butyl)phenyl]ethan-1-one, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.