missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-piperidone hydrate hydrochloride, 95%, Thermo Scientific Chemicals

236.08 SEK - 5442.08 SEK

Chemical Identifiers

CAS 50606-58-1
Molecular Formula C12H16ClNO
Molecular Weight (g/mol) 225.72
MDL Number MFCD00150096
InChI Key OVWSFXNSJDMRPV-UHFFFAOYSA-N
Synonym 1-benzyl-3-piperidone hydrochloride, 1-benzylpiperidin-3-one hydrochloride, 1-benzyl-3-piperidinone hydrochloride, 1-benzyl-3-piperidone hydrate hydrochloride, 1-benzylpiperidin-3-one hcl, 1-benzyl-3-piperidone hcl hydrate, 3-piperidinone, 1-phenylmethyl-, hydrochloride, 1-benzyl-3-piperidinone hydrate hydrochloride, n-benzyl-3-piperidone hydrochloride, pubchem7726Show MoreShow Less
PubChem CID 3084924
IUPAC Name 1-benzylpiperidin-3-one;hydrochloride
SMILES Cl.O=C1CCCN(CC2=CC=CC=C2)C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10637504
SDS View Documents
Thermo Scientific Acros
166920010
1 g Glass bottle
236.08 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
10515881
SDS View Documents
Thermo Scientific Acros
166920050
5 g Glass bottle N/A N/A N/A
10387661
SDS View Documents
Thermo Scientific
166920250
25 g Glass bottle N/A N/A N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Benzyl-3-piperidone hydrate hydrochloride, 95%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.