missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-4-hydroxypiperidine, 97%, Thermo Scientific™

129.00 SEK - 290.69 SEK

Chemical Identifiers

CAS 4727-72-4
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.27
MDL Number MFCD00006503
InChI Key BPPZXJZYCOETDA-UHFFFAOYSA-N
Synonym 1-benzyl-4-hydroxypiperidine, 1-benzyl-4-piperidinol, n-benzyl-4-hydroxypiperidine, 1-benzyl-piperidin-4-ol, 4-piperidinol, 1-phenylmethyl, n-benzyl-4-hydroxy piperidine, n-benzyl-4-piperidinol, 1-benzyl-4-hydroxy piperidine, 1-phenylmethyl piperidin-4-ol, 1-phenylmethyl-4-piperidinol
PubChem CID 78461
IUPAC Name 1-benzylpiperidin-4-ol
SMILES C1CN(CCC1O)CC2=CC=CC=C2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10667314
SDS View Documents
Thermo Scientific Acros
166830050
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10448910
SDS View Documents
Thermo Scientific Acros
166830250
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Benzyl-4-hydroxypiperidine, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.