missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2,3,5,6-tetrafluorobenzene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
1-Bromo-2,3,5,6-tetrafluorobenzene is used to produce 1,2,4,5-tetrafluoro-3-hept-1-ynyl-benzene. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromo-2,3,5,6-tetrafluorobenzene is used to produce 1,2,4,5-tetrafluoro-3-hept-1-ynyl-benzene. It is also used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
1-Bromo-2,3,5,6-tetrafluorobenzene is used to produce 1,2,4,5-tetrafluoro-3-hept-1-ynyl-benzene. It is also used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Specifications
Specifications
Density | 1.883 |
Boiling Point | 142°C to 144°C |
Flash Point | 53°C (129°F) |
Odor | Aromatic |
Refractive Index | 1.469 |
Quantity | 1 g |
UN Number | UN1993 |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 228.98 |
Percent Purity | 99% |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
1-Bromo-2,3,5,6-tetrafluorobenzene, 99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction