Learn More
1-Bromo-2-ethylbenzene, 98%, Thermo Scientific Chemicals
469.00 SEK - 6355.00 SEK
Chemical Identifiers
CAS | 1973-22-4 |
---|---|
Molecular Formula | C8H9Br |
Molecular Weight (g/mol) | 185.064 |
MDL Number | MFCD00000077 |
InChI Key | HVRUGFJYCAFAAN-UHFFFAOYSA-N |
PubChem CID | 16089 |
IUPAC Name | 1-bromo-2-ethylbenzene |
SMILES | CCC1=CC=CC=C1Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11475427
|
Thermo Scientific Alfa Aesar
B23449.06 |
5 g |
469.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485427
|
Thermo Scientific Alfa Aesar
B23449.14 |
25 g |
1845.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11495427
|
Thermo Scientific Alfa Aesar
B23449.22 |
100 g |
6355.00 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1973-22-4 | |
185.064 | |
HVRUGFJYCAFAAN-UHFFFAOYSA-N | |
1-bromo-2-ethylbenzene |
C8H9Br | |
MFCD00000077 | |
16089 | |
CCC1=CC=CC=C1Br |
Specifications
1973-22-4 | |
1.338 | |
71°C (159°F) | |
1.5485 | |
1929735 | |
HVRUGFJYCAFAAN-UHFFFAOYSA-N | |
1-bromo-2-ethylbenzene | |
16089 | |
98% |
-68°C | |
199°C to 200°C | |
C8H9Br | |
MFCD00000077 | |
o-bromoethylbenzene, benzene, 1-bromo-2-ethyl, 2-ethylbromobenzene, 1-ethyl-2-bromobenzene, 2-bromo-1-ethylbenzene, 1-bromo-2-ethyl-benzene, benzene,1-bromo-2-ethyl, o-ethylbromobenzene, 2-bromophenyl ethane, bromo-2-ethylbenzene | |
CCC1=CC=CC=C1Br | |
185.064 | |
185.06 | |
1-Bromo-2-ethylbenzene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'einecsNumber' : 217-823-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only