missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-pyrazole-5-boronic acid pinacol ester, 98%, Thermo Scientific™

508.00 SEK - 1239.00 SEK

Chemical Identifiers

CAS 847818-74-0
Molecular Formula C10H17BN2O2
Molecular Weight (g/mol) 208.07
MDL Number MFCD05861380
InChI Key HLXOVAMYQUFLPE-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-5-boronic acid pinacol ester, 1-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, 1-methylpyrazole-5-boronic acid pinacol ester, cbi-bb zero/005567, 1h-pyrazole, 1-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 1-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 4,4,5,5-tetramethyl-2-1-methylpyrazol-5-yl-1,3,2-dioxaborolane
PubChem CID 4912908
IUPAC Name 1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES CN1N=CC=C1B1OC(C)(C)C(C)(C)O1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11436170
SDS View Documents
Thermo Scientific Alfa Aesar
H27094.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15477655
SDS View Documents
Thermo Scientific Alfa Aesar
H27094.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester is used as a pharmaceutical intermediate, agrochemical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester, 98%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.