missing translation for 'onlineSavingsMsg'
Learn More
1-Methylimidazole-4-sulfonyl chloride, 95%, Thermo Scientific Chemicals Product Code.: 15406047

1-Methylimidazole-4-sulfonyl chloride, 95%, Thermo Scientific Chemicals

Product Code. 15406047
10 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 15406047

Brand: Thermo Scientific Alfa Aesar H33229.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137049-00-4
Molecular Formula C4H5ClN2O2S
Molecular Weight (g/mol) 180.61
MDL Number MFCD00068060
InChI Key KXUGUWTUFUWYRS-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-4-sulfonyl chloride, 1-methyl-1h-imidazole-4-sulphonyl chloride, 1h-imidazole-4-sulfonyl chloride, 1-methyl, 1-methylimidazole-4-sulfonylchloride, 1-methylimidazole-4-sulphonyl chloride, chloro 1-methylimidazol-4-yl sulfone, 2-methoxymethylpropenal, acmc-209cak, buttpark 27\06-11, 1-methylimidazol-4-sulfonylchlorideShow MoreShow Less
PubChem CID 2736887
IUPAC Name 1-methylimidazole-4-sulfonyl chloride
SMILES CN1C=NC(=C1)S(Cl)(=O)=O

Specifications

Melting Point 89°C to 91°C
Quantity 10 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 180.61
Percent Purity 95%
Chemical Name or Material 1-Methylimidazole-4-sulfonyl chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10175407 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33229.09,H33229.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methylimidazole-4-sulfonyl chloride, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.