missing translation for 'onlineSavingsMsg'
Learn More

1-Methylimidazole-4-sulfonyl chloride, 95%, Thermo Scientific Chemicals

381.00 SEK - 2457.05 SEK

Chemical Identifiers

CAS 137049-00-4
Molecular Formula C4H5ClN2O2S
Molecular Weight (g/mol) 180.61
MDL Number MFCD00068060
InChI Key KXUGUWTUFUWYRS-UHFFFAOYSA-N
Synonym 1-methyl-1h-imidazole-4-sulfonyl chloride, 1-methyl-1h-imidazole-4-sulphonyl chloride, 1h-imidazole-4-sulfonyl chloride, 1-methyl, 1-methylimidazole-4-sulfonylchloride, 1-methylimidazole-4-sulphonyl chloride, chloro 1-methylimidazol-4-yl sulfone, 2-methoxymethylpropenal, acmc-209cak, buttpark 27\06-11, 1-methylimidazol-4-sulfonylchloride
PubChem CID 2736887
IUPAC Name 1-methylimidazole-4-sulfonyl chloride
SMILES CN1C=NC(=C1)S(Cl)(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15496037
SDS View Documents
Thermo Scientific Alfa Aesar
H33229.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15406047
SDS View Documents
Thermo Scientific Alfa Aesar
H33229.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methylimidazole-4-sulfonyl chloride, 95%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.