missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-phenyl-1H-imidazole-2-carbaldehyde, Thermo Scientific™
975.00 SEK - 7905.00 SEK
Chemical Identifiers
CAS | 6002-15-9 |
---|---|
Molecular Formula | C10H8N2O |
Molecular Weight (g/mol) | 172.187 |
InChI Key | VJDRBJPZIHUDNE-UHFFFAOYSA-N |
Synonym | 1-phenyl-1h-imidazole-2-carbaldehyde, 1h-imidazole-2-carboxaldehyde, 1-phenyl, 1h-imidazole-2-carbaldehyde, 1-phenyl, 1-phenyl-2-formylimidazole, 1-phenylimidazole-2-carboxaldehyde |
PubChem CID | 6424701 |
IUPAC Name | 1-phenylimidazole-2-carbaldehyde |
SMILES | C1=CC=C(C=C1)N2C=CN=C2C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10133044
|
Thermo Scientific Maybridge
CC75904CB |
250 mg | Amber Glass Bottle |
975.00 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10052214
|
Thermo Scientific Maybridge
CC75904DA |
1 g | Amber Glass Bottle |
2725.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10602695
|
Thermo Scientific Maybridge
CC75904DE |
5 g | Amber Glass Bottle |
7905.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
6002-15-9 | |
172.187 | |
1-phenyl-1h-imidazole-2-carbaldehyde, 1h-imidazole-2-carboxaldehyde, 1-phenyl, 1h-imidazole-2-carbaldehyde, 1-phenyl, 1-phenyl-2-formylimidazole, 1-phenylimidazole-2-carboxaldehyde | |
1-phenylimidazole-2-carbaldehyde |
C10H8N2O | |
VJDRBJPZIHUDNE-UHFFFAOYSA-N | |
6424701 | |
C1=CC=C(C=C1)N2C=CN=C2C=O |
Specifications
6002-15-9 | |
1-phenyl-1h-imidazole-2-carbaldehyde, 1h-imidazole-2-carboxaldehyde, 1-phenyl, 1h-imidazole-2-carbaldehyde, 1-phenyl, 1-phenyl-2-formylimidazole, 1-phenylimidazole-2-carboxaldehyde | |
C1=CC=C(C=C1)N2C=CN=C2C=O | |
172.187 | |
172.19 | |
1-phenyl-1H-imidazole-2-carbaldehyde |
C10H8N2O | |
VJDRBJPZIHUDNE-UHFFFAOYSA-N | |
1-phenylimidazole-2-carbaldehyde | |
6424701 | |
97% |