Learn More
2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]-1,2-oxazinane, ≥97%, Thermo Scientific™
383.58 SEK - 511.82 SEK
Chemical Identifiers
CAS | 859833-22-0 |
---|---|
Molecular Formula | C18H28BNO2 |
Molecular Weight (g/mol) | 301.24 |
MDL Number | MFCD08271932 |
InChI Key | HJKVRMHVLINDRH-UHFFFAOYSA-N |
Synonym | 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperidine, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 4-1-piperidinylmethyl benzeneboronic acid pinacol ester, piperidine,1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, amtb120, 4-piperidinomethyl phenylboronic acid pinacol ester, 4-piperidinomethyl phenylboronic acid, pinacol ester, 4-piperidin-1-ylmethyl phenyl boronic acid pinacol esterShow More |
PubChem CID | 18525779 |
IUPAC Name | 1-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperidine |
SMILES | CC1(C)OB(OC1(C)C)C1=CC=C(CN2CCCCC2)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11360627
|
Thermo Scientific Maybridge
CC45539CB |
250 mg | Amber Glass Bottle |
383.58 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10307304
|
Thermo Scientific Maybridge
CC45539DA |
1 g | Amber Glass Bottle |
511.82 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
859833-22-0 | |
301.24 | |
HJKVRMHVLINDRH-UHFFFAOYSA-N | |
18525779 | |
CC1(C)OB(OC1(C)C)C1=CC=C(CN2CCCCC2)C=C1 |
C18H28BNO2 | |
MFCD08271932 | |
1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperidine, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 4-1-piperidinylmethyl benzeneboronic acid pinacol ester, piperidine,1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, amtb120, 4-piperidinomethyl phenylboronic acid pinacol ester, 4-piperidinomethyl phenylboronic acid, pinacol ester, 4-piperidin-1-ylmethyl phenyl boronic acid pinacol esterShow More | |
1-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperidine |
Specifications
859833-22-0 | |
100.0 | |
C18H28BNO2 | |
1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl piperidine, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl piperidine, 4-1-piperidinylmethyl benzeneboronic acid pinacol ester, piperidine,1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl, amtb120, 4-piperidinomethyl phenylboronic acid pinacol ester, 4-piperidinomethyl phenylboronic acid, pinacol ester, 4-piperidin-1-ylmethyl phenyl boronic acid pinacol ester | |
CC1(C)OB(OC1(C)C)C1=CC=C(CN2CCCCC2)C=C1 | |
301.24 | |
301.24 | |
1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperidine |
96.5 | |
97% | |
MFCD08271932 | |
HJKVRMHVLINDRH-UHFFFAOYSA-N | |
1-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperidine | |
18525779 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
Your input is important to us. Please complete this form to provide feedback related to the content on this product.