missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trimethylbenzeneboronic acid neopentyl glycol ester, 99%, Thermo Scientific™
461.00 SEK - 1145.00 SEK
Chemical Identifiers
CAS | 214360-78-8 |
---|---|
Molecular Formula | C14H21BO2 |
Molecular Weight (g/mol) | 232.13 |
MDL Number | MFCD01318178 |
InChI Key | ITLDRNGUSKUMRG-UHFFFAOYSA-N |
Synonym | 2,4,6-trimethylbenzeneboronic acid neopentyl glycol ester, 5,5-dimethyl-2-2,4,6-trimethylphenyl-1,3,2-dioxaborinane, 2-mesityl-5,5-dimethyl-1,3,2-dioxaborinane, 2,4,6-trimethylbenzeneboronic acid neopentyl glycol cyclic ester, mesitylboronic acid 2,2-dimethyltrimethylene ester, 2,4,6-trimethyl-benzeneboronic acid neopentyl glycol cyclic ester, 2,4,6-trimethylphenylboronic acid neopentyl glycol ester, mesityleneboronic acid 2,2-dimethyltrimethylene cyclic ester |
PubChem CID | 11436188 |
IUPAC Name | 5,5-dimethyl-2-(2,4,6-trimethylphenyl)-1,3,2-dioxaborinane |
SMILES | B1(OCC(CO1)(C)C)C2=C(C=C(C=C2C)C)C |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
214360-78-8 | |
232.13 | |
ITLDRNGUSKUMRG-UHFFFAOYSA-N | |
11436188 | |
B1(OCC(CO1)(C)C)C2=C(C=C(C=C2C)C)C |
C14H21BO2 | |
MFCD01318178 | |
2,4,6-trimethylbenzeneboronic acid neopentyl glycol ester, 5,5-dimethyl-2-2,4,6-trimethylphenyl-1,3,2-dioxaborinane, 2-mesityl-5,5-dimethyl-1,3,2-dioxaborinane, 2,4,6-trimethylbenzeneboronic acid neopentyl glycol cyclic ester, mesitylboronic acid 2,2-dimethyltrimethylene ester, 2,4,6-trimethyl-benzeneboronic acid neopentyl glycol cyclic ester, 2,4,6-trimethylphenylboronic acid neopentyl glycol ester, mesityleneboronic acid 2,2-dimethyltrimethylene cyclic ester | |
5,5-dimethyl-2-(2,4,6-trimethylphenyl)-1,3,2-dioxaborinane |
Specifications
214360-78-8 | |
C14H21BO2 | |
8689636 | |
ITLDRNGUSKUMRG-UHFFFAOYSA-N | |
5,5-dimethyl-2-(2,4,6-trimethylphenyl)-1,3,2-dioxaborinane | |
11436188 | |
99% |
46°C to 48°C | |
MFCD01318178 | |
2,4,6-trimethylbenzeneboronic acid neopentyl glycol ester, 5,5-dimethyl-2-2,4,6-trimethylphenyl-1,3,2-dioxaborinane, 2-mesityl-5,5-dimethyl-1,3,2-dioxaborinane, 2,4,6-trimethylbenzeneboronic acid neopentyl glycol cyclic ester, mesitylboronic acid 2,2-dimethyltrimethylene ester, 2,4,6-trimethyl-benzeneboronic acid neopentyl glycol cyclic ester, 2,4,6-trimethylphenylboronic acid neopentyl glycol ester, mesityleneboronic acid 2,2-dimethyltrimethylene cyclic ester | |
B1(OCC(CO1)(C)C)C2=C(C=C(C=C2C)C)C | |
232.13 | |
232.13 | |
2,4,6-Trimethylbenzeneboronic acid neopentyl glycol ester |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2,4,6-Trimethylbenzeneboronic acid neopentyl glycol ester, 99%, Thermo Scientific™