missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dimethoxypyridine-3-boronic acid, 97%, Thermo Scientific™
1109.00 SEK - 4663.00 SEK
Chemical Identifiers
CAS | 221006-70-8 |
---|---|
Molecular Formula | C7H10BNO4 |
Molecular Weight (g/mol) | 182.97 |
InChI Key | ADGHSWFUZUADDH-UHFFFAOYSA-N |
Synonym | 2,6-dimethoxypyridine-3-boronic acid, 2,6-dimethoxy-3-pyridineboronic acid, 2,6-dimethoxypyridin-3-yl boronic acid, 2,6-dimethoxypyridin-3-yl-3-boronic acid, 3-borono-2,6-dimethoxypyridine, 2,6-dimethoxy-3-pyridylboronic acid, 2,6-dimethoxypyridine-5-boronic acid, boronic acid, 2,6-dimethoxy-3-pyridinyl, 2,6-dimethoxy-3-pyridyl boronic acid |
PubChem CID | 2762707 |
IUPAC Name | (2,6-dimethoxypyridin-3-yl)boronic acid |
SMILES | B(C1=C(N=C(C=C1)OC)OC)(O)O |
Description
Chemical Identifiers
221006-70-8 | |
182.97 | |
2,6-dimethoxypyridine-3-boronic acid, 2,6-dimethoxy-3-pyridineboronic acid, 2,6-dimethoxypyridin-3-yl boronic acid, 2,6-dimethoxypyridin-3-yl-3-boronic acid, 3-borono-2,6-dimethoxypyridine, 2,6-dimethoxy-3-pyridylboronic acid, 2,6-dimethoxypyridine-5-boronic acid, boronic acid, 2,6-dimethoxy-3-pyridinyl, 2,6-dimethoxy-3-pyridyl boronic acid | |
(2,6-dimethoxypyridin-3-yl)boronic acid |
C7H10BNO4 | |
ADGHSWFUZUADDH-UHFFFAOYSA-N | |
2762707 | |
B(C1=C(N=C(C=C1)OC)OC)(O)O |
Specifications
221006-70-8 | |
96% min. (HPLC) | |
C7H10BNO4 | |
ADGHSWFUZUADDH-UHFFFAOYSA-N | |
(2,6-dimethoxypyridin-3-yl)boronic acid | |
2762707 | |
97% |
108°C to 109°C | |
Glass Bottle | |
2,6-dimethoxypyridine-3-boronic acid, 2,6-dimethoxy-3-pyridineboronic acid, 2,6-dimethoxypyridin-3-yl boronic acid, 2,6-dimethoxypyridin-3-yl-3-boronic acid, 3-borono-2,6-dimethoxypyridine, 2,6-dimethoxy-3-pyridylboronic acid, 2,6-dimethoxypyridine-5-boronic acid, boronic acid, 2,6-dimethoxy-3-pyridinyl, 2,6-dimethoxy-3-pyridyl boronic acid | |
B(C1=C(N=C(C=C1)OC)OC)(O)O | |
182.97 | |
182.97 | |
2, 6-Dimethoxypyridine-3-boronic acid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protectio
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2,6-Dimethoxypyridine-3-boronic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2,6-Dimethoxypyridine-3-boronic acid, 97%, Thermo Scientific™