missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-nitro-4-picoline, 98%, Thermo Scientific™

185.00 SEK - 1822.40 SEK

Chemical Identifiers

CAS 6635-86-5
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00006315
InChI Key IKMZGACFMXZAAT-UHFFFAOYSA-N
Synonym 2-amino-4-methyl-3-nitropyridine, 2-amino-3-nitro-4-picoline, 2-pyridinamine, 4-methyl-3-nitro, 2-amino-3-nitro-4-methylpyridine, 4-methyl-3-nitro-pyridin-2-ylamine, 4-methyl-3-nitro-2-pyridylamine, 4-methyl-3-nitro-2-pyridinamine, 4-methyl-3-nitro-2-aminopyridine, 4-methyl-3-nitropyridin-2-amin, 4mrz
PubChem CID 243166
IUPAC Name 4-methyl-3-nitropyridin-2-amine
SMILES CC1=C(C(=NC=C1)N)[N+](=O)[O-]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10123641
View Documents
Thermo Scientific Acros
319550010
1 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10043610
View Documents
Thermo Scientific Acros
319550050
5 g Glass bottle
481.63 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
10701545
View Documents
Thermo Scientific Acros
319550250
25 g Glass bottle
1822.40 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

6635-86-5
153.14
IKMZGACFMXZAAT-UHFFFAOYSA-N
243166
CC1=C(C(=NC=C1)N)[N+](=O)[O-]
C6H7N3O2
MFCD00006315
2-amino-4-methyl-3-nitropyridine, 2-amino-3-nitro-4-picoline, 2-pyridinamine, 4-methyl-3-nitro, 2-amino-3-nitro-4-methylpyridine, 4-methyl-3-nitro-pyridin-2-ylamine, 4-methyl-3-nitro-2-pyridylamine, 4-methyl-3-nitro-2-pyridinamine, 4-methyl-3-nitro-2-aminopyridine, 4-methyl-3-nitropyridin-2-amin, 4mrz
4-methyl-3-nitropyridin-2-amine

Specifications

6635-86-5
100.0
Yellow
97.5% min. (GC)
MFCD00006315
IKMZGACFMXZAAT-UHFFFAOYSA-N
4-methyl-3-nitropyridin-2-amine
243166
98%
2-Amino-3-nitro-4-picoline
97.5
136°C to 141°C
Authentic
C6H7N3O2
2-amino-4-methyl-3-nitropyridine, 2-amino-3-nitro-4-picoline, 2-pyridinamine, 4-methyl-3-nitro, 2-amino-3-nitro-4-methylpyridine, 4-methyl-3-nitro-pyridin-2-ylamine, 4-methyl-3-nitro-2-pyridylamine, 4-methyl-3-nitro-2-pyridinamine, 4-methyl-3-nitro-2-aminopyridine, 4-methyl-3-nitropyridin-2-amin, 4mrz
CC1=C(C(=NC=C1)N)[N+](=O)[O-]
153.14
153.14
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only