missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(2-chloro-3-pyridyl)pyrimidine, 97%, Thermo Scientific™

2071.00 SEK - 7855.00 SEK

Chemical Identifiers

CAS 870221-49-1
Molecular Formula C9H7ClN4
Molecular Weight (g/mol) 206.633
MDL Number MFCD18157687
InChI Key DKCVVHMYFZBDAK-UHFFFAOYSA-N
PubChem CID 53396335
IUPAC Name 4-(2-chloropyridin-3-yl)pyrimidin-2-amine
SMILES C1=CC(=C(N=C1)Cl)C2=NC(=NC=C2)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15411098
View Documents
Thermo Scientific Alfa Aesar
H54957.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15401098
View Documents
Thermo Scientific Alfa Aesar
H54957.03
1 g
7855.00 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as primary and secondary intermediate.

Notes
Incompatible with strong oxidizing agents, bases, acids. Keep in room temperature.
Specifications

Chemical Identifiers

870221-49-1
206.633
DKCVVHMYFZBDAK-UHFFFAOYSA-N
4-(2-chloropyridin-3-yl)pyrimidin-2-amine
C9H7ClN4
MFCD18157687
53396335
C1=CC(=C(N=C1)Cl)C2=NC(=NC=C2)N

Specifications

870221-49-1
MFCD18157687
4-2-chloropyridin-3-yl pyrimidin-2-amine, chloropyridinylpyrimidinamine, 2-pyrimidinamine,4-2-chloro-3-pyridinyl, 2-pyrimidinamine, 4-2-chloro-3-pyridinyl
C1=CC(=C(N=C1)Cl)C2=NC(=NC=C2)N
206.633
206.63
2-Amino-4-(2-chloro-3-pyridyl)pyrimidine
C9H7ClN4
14484335
DKCVVHMYFZBDAK-UHFFFAOYSA-N
4-(2-chloropyridin-3-yl)pyrimidin-2-amine
53396335
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications