Learn More
2-Butynal diethyl acetal, 98%, Thermo Scientific Chemicals
444.00 SEK - 1070.00 SEK
Chemical Identifiers
CAS | 2806-97-5 |
---|---|
Molecular Formula | C8H14O2 |
Molecular Weight (g/mol) | 142.20 |
MDL Number | MFCD00009274 |
InChI Key | FKKVKKSEVMQYER-UHFFFAOYSA-N |
Synonym | 2-butynal diethyl acetal, 2-butyn-1-al diethyl acetal, 2-butynyl aldehyde diethyl acetal, 2-butyne, 1,1-diethoxy, 1,1-diethoxy-but-2-yne, 1-diethoxybut-2-yne, acmc-20ao12, tetrolaldehyde diethyl acetal, 1,1-diethoxy-2-butyne # |
PubChem CID | 137721 |
IUPAC Name | 1,1-diethoxybut-2-yne |
SMILES | CCOC(OCC)C#CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11327625
|
Thermo Scientific Alfa Aesar
L05872.03 |
1 g |
444.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337625
|
Thermo Scientific Alfa Aesar
L05872.06 |
5 g |
1070.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Butynal diethyl acetal is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Butynal diethyl acetal is used as pharmaceutical intermediates.
Solubility
Not miscible in water.
Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
Chemical Identifiers
2806-97-5 | |
142.20 | |
FKKVKKSEVMQYER-UHFFFAOYSA-N | |
137721 | |
CCOC(OCC)C#CC |
C8H14O2 | |
MFCD00009274 | |
2-butynal diethyl acetal, 2-butyn-1-al diethyl acetal, 2-butynyl aldehyde diethyl acetal, 2-butyne, 1,1-diethoxy, 1,1-diethoxy-but-2-yne, 1-diethoxybut-2-yne, acmc-20ao12, tetrolaldehyde diethyl acetal, 1,1-diethoxy-2-butyne # | |
1,1-diethoxybut-2-yne |
Specifications
2806-97-5 | |
65°C to 67°C (12 mmHg) | |
Recognizable | |
1.426 | |
UN1993 | |
2-butynal diethyl acetal, 2-butyn-1-al diethyl acetal, 2-butynyl aldehyde diethyl acetal, 2-butyne, 1,1-diethoxy, 1,1-diethoxy-but-2-yne, 1-diethoxybut-2-yne, acmc-20ao12, tetrolaldehyde diethyl acetal, 1,1-diethoxy-2-butyne # | |
FKKVKKSEVMQYER-UHFFFAOYSA-N | |
1,1-diethoxybut-2-yne | |
137721 | |
98% |
0.898 | |
56°C (133°F) | |
C8H14O2 | |
MFCD00009274 | |
1702075 | |
Not miscible in water. | |
CCOC(OCC)C#CC | |
142.20 | |
142.2 | |
2-Butynal diethyl acetal |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.