missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-1-naphthoic acid, 98%, Thermo Scientific™

593.00 SEK - 1395.00 SEK

Chemical Identifiers

CAS 2224-00-2
Molecular Formula C13H11O3
Molecular Weight (g/mol) 215.23
MDL Number MFCD00004008
InChI Key MYFBSSDLYGWAHH-UHFFFAOYSA-M
Synonym 2-ethoxy-1-naphthoic acid, 2-ethoxynaphthoic acid, 1-naphthalenecarboxylic acid, 2-ethoxy, 2-ethoxy-1-naphthoicacid, 2-ethoxynaphthalenecarboxylic acid, acmc-209ftv, rarechem al be 0566, timtec-bb sbb005771, 2-ethoxynaphtoic acid
PubChem CID 75210
IUPAC Name 2-ethoxynaphthalene-1-carboxylic acid
SMILES CCOC1=CC=C2C=CC=CC2=C1C([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11495287
View Documents
Thermo Scientific Alfa Aesar
B22665.14
25 g
593.00 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
11405297
View Documents
Thermo Scientific Alfa Aesar
B22665.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2224-00-2
215.23
MYFBSSDLYGWAHH-UHFFFAOYSA-M
75210
CCOC1=CC=C2C=CC=CC2=C1C([O-])=O
C13H11O3
MFCD00004008
2-ethoxy-1-naphthoic acid, 2-ethoxynaphthoic acid, 1-naphthalenecarboxylic acid, 2-ethoxy, 2-ethoxy-1-naphthoicacid, 2-ethoxynaphthalenecarboxylic acid, acmc-209ftv, rarechem al be 0566, timtec-bb sbb005771, 2-ethoxynaphtoic acid
2-ethoxynaphthalene-1-carboxylic acid

Specifications

2224-00-2
C13H11O3
2695612
MYFBSSDLYGWAHH-UHFFFAOYSA-M
2-ethoxynaphthalene-1-carboxylic acid
75210
98%
143°C to 145°C
MFCD00004008
2-ethoxy-1-naphthoic acid, 2-ethoxynaphthoic acid, 1-naphthalenecarboxylic acid, 2-ethoxy, 2-ethoxy-1-naphthoicacid, 2-ethoxynaphthalenecarboxylic acid, acmc-209ftv, rarechem al be 0566, timtec-bb sbb005771, 2-ethoxynaphtoic acid
CCOC1=CC=C2C=CC=CC2=C1C([O-])=O
215.23
216.24
2-Ethoxy-1-naphthoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 218-745-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.