Learn More
2-Fluoro-4-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
2265.00 SEK - 7155.00 SEK
Chemical Identifiers
CAS | 1259022-70-2 |
---|---|
Molecular Formula | C15H20BFO4 |
Molecular Weight (g/mol) | 294.13 |
MDL Number | MFCD18427635 |
InChI Key | ZFLOTGNTGSGUGD-UHFFFAOYSA-N |
Synonym | methyl 2-3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 2-fluoro-4-methoxycarbonylmethyl benzeneboronic acid pinacol ester, methyl 2-3-fluoro-4-4,4,5 5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate |
PubChem CID | 59627674 |
IUPAC Name | methyl 2-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate |
SMILES | COC(=O)CC1=CC(F)=C(C=C1)B1OC(C)(C)C(C)(C)O1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1259022-70-2 | |
294.13 | |
ZFLOTGNTGSGUGD-UHFFFAOYSA-N | |
59627674 | |
COC(=O)CC1=CC(F)=C(C=C1)B1OC(C)(C)C(C)(C)O1 |
C15H20BFO4 | |
MFCD18427635 | |
methyl 2-3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 2-fluoro-4-methoxycarbonylmethyl benzeneboronic acid pinacol ester, methyl 2-3-fluoro-4-4,4,5 5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate | |
methyl 2-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate |
Specifications
1259022-70-2 | |
MFCD18427635 | |
ZFLOTGNTGSGUGD-UHFFFAOYSA-N | |
methyl 2-[3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate | |
59627674 | |
96% |
C15H20BFO4 | |
methyl 2-3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate, 2-fluoro-4-methoxycarbonylmethyl benzeneboronic acid pinacol ester, methyl 2-3-fluoro-4-4,4,5 5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetate | |
COC(=O)CC1=CC(F)=C(C=C1)B1OC(C)(C)C(C)(C)O1 | |
294.13 | |
294.13 | |
2-Fluoro-4-(methoxycarbonylmethyl)benzeneboronic acid pinacol ester |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.