missing translation for 'onlineSavingsMsg'
Learn More

2-Formylbenzenesulfonic acid sodium salt, 90%, Tech., Thermo Scientific Chemicals

1437.07 SEK - 4824.44 SEK

Chemical Identifiers

CAS 1008-72-6
Molecular Formula C7H5NaO4S
Molecular Weight (g/mol) 208.16
MDL Number MFCD00007478
InChI Key ADPUQRRLAAPXGT-UHFFFAOYSA-M
Synonym sodium 2-formylbenzenesulfonate, 2-formylbenzenesulfonic acid sodium salt, sodium o-formylbenzenesulfonate, benzenesulfonic acid, 2-formyl-, sodium salt, unii-lh8xh59068, 2-sulfobenzaldehyde sodium salt, sodium benzaldehyde-2-sulfonate, benzaldehyde-2-sulfonic acid sodium salt, o-formylbenzenesulfonic acid sodium salt, sodium 2-formylbenzene-1-sulfonate
PubChem CID 3794540
IUPAC Name sodium;2-formylbenzenesulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=CC=C1C=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10175130
SDS View Documents
Thermo Scientific Acros
219325000
500 g Plastic bottle
1437.07 SEK
500g
Please to purchase this item. Need a web account? Register with us today!
10043863
SDS View Documents
Thermo Scientific Acros
219320025
2.5 kg Plastic bottle
4824.44 SEK
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

AIR SENSITIVE

missing translation for 'einecsNumber' : 213-758-4

missing translation for 'tsca' : TSCA

Product Identifier
  • 2-Formylbenzenesulfonic acid sodium salt

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Formylbenzenesulfonic acid sodium salt, 90%, Tech., Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.