missing translation for 'onlineSavingsMsg'
Learn More
2-Hydroxy-3-methoxybenzaldehyde, 99%, Thermo Scientific Chemicals Product Code.: 11457607

2-Hydroxy-3-methoxybenzaldehyde, 99%, Thermo Scientific Chemicals

Product Code. 11457607
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11457607

Brand: Thermo Scientific Alfa Aesar A15672.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

o-Vanillin has been used to study the solvent-free reaction between o-vanillin and p-toluidine using NMR, DSC and XRD analysis. It was used in the synthesis of new ligand for Fe(III) and Al(lII). It is also used in the study of mutagenesis and as a synthetic precursor for pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Vanillin has been used to study the solvent-free reaction between o-vanillin and p-toluidine using NMR, DSC and XRD analysis. It was used in the synthesis of new ligand for Fe(III) and Al(lII). It is also used in the study of mutagenesis and as a synthetic precursor for pharmaceuticals.

Solubility
Solubility in methanol is almost transparent. Soluble in alcohol, methanol and dioxane (1g in 10 mL). Insoluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148-53-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00003322
InChI Key JJVNINGBHGBWJH-UHFFFAOYSA-N
Synonym 3-methoxysalicylaldehyde, o-vanillin, 6-formylguaiacol, 2-vanillin, o-vanilline, benzaldehyde, 2-hydroxy-3-methoxy, orthovanilline, 2-hydroxy-m-anisaldehyde, 6-formyl-2-methoxyphenol, 3-methoxy-2-hydroxybenzaldehydeShow MoreShow Less
PubChem CID 8991
ChEBI CHEBI:78339
IUPAC Name 2-hydroxy-3-methoxybenzaldehyde
SMILES COC1=CC=CC(C=O)=C1O

Specifications

Melting Point 40°C to 42°C
Boiling Point 265°C to 266°C
Quantity 25 g
Beilstein 471913
Sensitivity Air Sensitive
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material 2-Hydroxy-3-methoxybenzaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3082132A Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberA15672.22,A15672.36,A15672.14
Lot Number2795178AA Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberA15672.22,A15672.36,A15672.14
Lot Number10251078 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA15672.22,A15672.14,A15672.36
Lot Number10218801 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberA15672.22,A15672.14,A15672.36
Lot Number10248640 Certificate TypeCertificate of Analysis Date22/11/2023 Catalog NumberA15672.22,A15672.14,A15672.36
compliance-icons
Product Identifier
  • 2-Hydroxy-3-methoxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2-Hydroxy-3-methoxybenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Hydroxy-3-methoxybenzaldehyde, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.