missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2-Methyl-5-nitroimidazole, 99%
404.71 SEK - 1295.00 SEK
Chemical Identifiers
CAS | 88054-22-2 |
---|---|
Molecular Formula | C4H5N3O2 |
Molecular Weight (g/mol) | 127.10 |
MDL Number | MFCD00005191 |
InChI Key | FFYTTYVSDVWNMY-UHFFFAOYSA-N |
Synonym | 2-methyl-5-nitroimidazole, 2-methyl-4-nitroimidazole, 2-methyl-4-nitro-1h-imidazole, menidazole, 1h-imidazole, 2-methyl-4-nitro, 1h-imidazole, 2-methyl-5-nitro, 2-methyl-4 5-nitroimidazole, imidazole, 2-methyl-4-nitro, unii-24ag2ww15w, imidazole, 2-methyl-5-nitro |
PubChem CID | 12760 |
IUPAC Name | 2-methyl-5-nitro-1H-imidazole |
SMILES | CC1=NC=C(N1)[N+]([O-])=O |
Description
Applications2-Methyl-4(5)-nitroimidazole was used to study the mechanism of both the alkaline and acidic hydrolysis of tinidazole. It is mainly used to synthesize intermediate of medicines such as Metronidazole and dimetridazole.
Solubility
Slightly soluble in ethanol, easily soluble in diluted acid and diluted alkali.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Identifiants chimiques
88054-22-2 | |
127.10 | |
FFYTTYVSDVWNMY-UHFFFAOYSA-N | |
12760 | |
CC1=NC=C(N1)[N+]([O-])=O |
C4H5N3O2 | |
MFCD00005191 | |
2-methyl-5-nitroimidazole, 2-methyl-4-nitroimidazole, 2-methyl-4-nitro-1h-imidazole, menidazole, 1h-imidazole, 2-methyl-4-nitro, 1h-imidazole, 2-methyl-5-nitro, 2-methyl-4 5-nitroimidazole, imidazole, 2-methyl-4-nitro, unii-24ag2ww15w, imidazole, 2-methyl-5-nitro | |
2-methyl-5-nitro-1H-imidazole |
Spécification
88054-22-2 | |
1.426g/mL | |
C4H5N3O2 | |
4032 | |
FFYTTYVSDVWNMY-UHFFFAOYSA-N | |
2-methyl-5-nitro-1H-imidazole | |
12760 | |
99% | |
2-Methyl-5-nitroimidazole |
251°C to 255°C | |
399°C | |
MFCD00005191 | |
2-methyl-5-nitroimidazole, 2-methyl-4-nitroimidazole, 2-methyl-4-nitro-1h-imidazole, menidazole, 1h-imidazole, 2-methyl-4-nitro, 1h-imidazole, 2-methyl-5-nitro, 2-methyl-4 5-nitroimidazole, imidazole, 2-methyl-4-nitro, unii-24ag2ww15w, imidazole, 2-methyl-5-nitro | |
CC1=NC=C(N1)[N+]([O-])=O | |
127.10 | |
127.1 | |
Solid |
Sécurité et traitement
GHS H Statement
H301
Toxic if swallowed.
GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger