missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophene-2-carboxamide, 99%, Thermo Scientific Chemicals

725.00 SEK - 2000.00 SEK

Chemical Identifiers

CAS 78031-18-2
Molecular Formula C5H4BrNOS
Molecular Weight (g/mol) 206.06
MDL Number MFCD00173835
InChI Key KWXUWNXTRUZHNQ-UHFFFAOYSA-N
Synonym maybridge1_007895, 3-bromo-2-carboxamidothiophene, 2-thiophenecarboxamide,3-bromo, 3-bromo-thiophene-2-carboxamide, 3-bromo-2-thiophenecarboxamide #, buttpark 180\02-08, 3-bromo thiophene-2-carboxamide
PubChem CID 605721
IUPAC Name 3-bromothiophene-2-carboxamide
SMILES NC(=O)C1=C(Br)C=CS1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11312907
SDS View Documents
Thermo Scientific Alfa Aesar
L16138.MD
250 mg N/A N/A N/A
11302907
SDS View Documents
Thermo Scientific Alfa Aesar
L16138.03
1 g
2000.00 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

3-Bromothiophene-2-carboxamide is used in Stille cross-coupling conditions were used to couple the resin-bound peptide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromothiophene-2-carboxamide is used in Stille cross-coupling conditions were used to couple the resin-bound peptide.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromothiophene-2-carboxamide, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.