Learn More
3-chlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
264.57 SEK - 3501.42 SEK
Chemical Identifiers
CAS | 2888-06-4 |
---|---|
Molecular Formula | C6H4Cl2O2S |
Molecular Weight (g/mol) | 211.07 |
MDL Number | MFCD00051697 |
InChI Key | OINWZUJVEXUHCC-UHFFFAOYSA-N |
Synonym | 3-chlorobenzene-1-sulfonyl chloride, 3-chlorobenzenesulphonyl chloride, 3-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 3-chloro, m-chlorobenzenesulfonyl chloride, 3-chloro-benzenesulfonyl chloride, 3-chlorophenylsulfonyl chloride, pubchem5076, m-chlorbenzolsulfochlorid, acmc-1cj4bShow More |
PubChem CID | 17909 |
IUPAC Name | 3-chlorobenzenesulfonyl chloride |
SMILES | C1=CC(=CC(=C1)Cl)S(=O)(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10539882
|
Thermo Scientific Acros
356480010 |
1 g | Glass bottle | N/A | ||||||
10056093
|
Thermo Scientific Acros
356480050 |
5 g | Glass bottle | N/A | ||||||
10219593
|
Thermo Scientific Acros
356480250 |
25 g | Glass bottle |
3501.42 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2888-06-4 | |
211.07 | |
OINWZUJVEXUHCC-UHFFFAOYSA-N | |
17909 | |
C1=CC(=CC(=C1)Cl)S(=O)(=O)Cl |
C6H4Cl2O2S | |
MFCD00051697 | |
3-chlorobenzene-1-sulfonyl chloride, 3-chlorobenzenesulphonyl chloride, 3-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 3-chloro, m-chlorobenzenesulfonyl chloride, 3-chloro-benzenesulfonyl chloride, 3-chlorophenylsulfonyl chloride, pubchem5076, m-chlorbenzolsulfochlorid, acmc-1cj4bShow More | |
3-chlorobenzenesulfonyl chloride |
Specifications
2888-06-4 | |
100.0 | |
1.4990g/mL | |
Authentic | |
C6H4Cl2O2S | |
1.5670 to 1.569 | |
1.499 | |
OINWZUJVEXUHCC-UHFFFAOYSA-N | |
3-chlorobenzenesulfonyl chloride | |
17909 | |
98% | |
3-chlorobenzenesulfonyl chloride |
97.5 | |
Yellow | |
102°C (0.9 mmHg) | |
98% | |
ClC6H4SO2Cl | |
MFCD00051697 | |
3-chlorobenzene-1-sulfonyl chloride, 3-chlorobenzenesulphonyl chloride, 3-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 3-chloro, m-chlorobenzenesulfonyl chloride, 3-chloro-benzenesulfonyl chloride, 3-chlorophenylsulfonyl chloride, pubchem5076, m-chlorbenzolsulfochlorid, acmc-1cj4b | |
C1=CC(=CC(=C1)Cl)S(=O)(=O)Cl | |
211.07 | |
211.07 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 220-753-
- 3-Chlorobenzenesulfonyl chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.