Learn More
3-Fluoro-5-(pentafluorothio)benzylamine, 97%, Thermo Scientific™
949.00 SEK - 3133.00 SEK
Chemische Identifikatoren
CAS | 1240257-73-1 |
---|---|
Molecular Formula | C7H7F6NS |
Molecular Weight (g/mol) | 251.19 |
MDL Number | MFCD16652463 |
InChI Key | FKGCFDAJSOIINF-UHFFFAOYSA-N |
Synonym | 3-fluoro-5-pentafluorosulfur benzylamine, 1-3-fluoro-5-pentafluoro-??-sulfanyl phenyl methanamine, 3-fluoro-5-pentafluorothio benzylamine |
PubChem CID | 66523547 |
IUPAC Name | [3-fluoro-5-(pentafluoro-$l^{6}-sulfanyl)phenyl]methanamine |
SMILES | NCC1=CC(F)=CC(=C1)S(F)(F)(F)(F)F |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | ||||||
15497147
|
Thermo Scientific Alfa Aesar
H34213.MD |
250 mg | N/A | N/A | N/A | |||||
15487147
|
Thermo Scientific Alfa Aesar
H34213.03 |
1 g | N/A | N/A | N/A | |||||
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemische Identifikatoren
1240257-73-1 | |
251.19 | |
FKGCFDAJSOIINF-UHFFFAOYSA-N | |
66523547 | |
NCC1=CC(F)=CC(=C1)S(F)(F)(F)(F)F |
C7H7F6NS | |
MFCD16652463 | |
3-fluoro-5-pentafluorosulfur benzylamine, 1-3-fluoro-5-pentafluoro-??-sulfanyl phenyl methanamine, 3-fluoro-5-pentafluorothio benzylamine | |
[3-fluoro-5-(pentafluoro-$l^{6}-sulfanyl)phenyl]methanamine |
Spezifikation
1240257-73-1 | |
MFCD16652463 | |
Air sensitive | |
FKGCFDAJSOIINF-UHFFFAOYSA-N | |
[3-fluoro-5-(pentafluoro-$l^{6}-sulfanyl)phenyl]methanamine | |
66523547 | |
97% |
C7H7F6NS | |
UN2735 | |
3-fluoro-5-pentafluorosulfur benzylamine, 1-3-fluoro-5-pentafluoro-??-sulfanyl phenyl methanamine, 3-fluoro-5-pentafluorothio benzylamine | |
NCC1=CC(F)=CC(=C1)S(F)(F)(F)(F)F | |
251.19 | |
251.19 | |
3-Fluoro-5-(pentafluorothio)benzylamine |
Sicherheit und Handhabung
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.