missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Fluorobenzoyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals

619.00 SEK - 7339.00 SEK

Chemical Identifiers

CAS 25519-78-2
Molecular Formula C12H15ClFNO
Molecular Weight (g/mol) 243.71
MDL Number MFCD00044912
InChI Key GPKDBZQZPNOBGM-UHFFFAOYSA-N
Synonym 4-4-fluorobenzoyl piperidine hydrochloride, 4-fluorophenyl piperidin-4-yl methanone hydrochloride, 4-fluorophenyl 4-piperidinyl ketone hydrochloride, p-fluorophenyl 4-piperidyl ketone hydrochloride, 4-fluorophenyl 4-piperidyl methanone hydrochloride, 4-fluoro-phenyl-piperidin-4-yl-methanone hydrochloride, 4-4-fluorobenzoyl piperidine hcl, 4-p-fluorobenzoyl piperidine hydrochloride, 4-4'-fluorobenzoyl piperidine hydrochloride, methanone, 4-fluorophenyl-4-piperidinyl-, hydrochloride
PubChem CID 3084438
IUPAC Name (4-fluorophenyl)-piperidin-4-ylmethanone;hydrochloride
SMILES [H+].[Cl-].FC1=CC=C(C=C1)C(=O)C1CCNCC1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11446366
SDS View Documents
Thermo Scientific Alfa Aesar
B23142.03
1 g
619.00 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
11456366
SDS View Documents
Thermo Scientific Alfa Aesar
B23142.06
5 g
2300.00 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
11466366
SDS View Documents
Thermo Scientific Alfa Aesar
B23142.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 247-070-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(4-Fluorobenzoyl)piperidine hydrochloride, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.