missing translation for 'onlineSavingsMsg'
Learn More
4-Bromo-2-methylbenzoic acid, 98+%, Thermo Scientific Chemicals Product Code.: 15468809

4-Bromo-2-methylbenzoic acid, 98+%, Thermo Scientific Chemicals

Product Code. 15468809
100 g
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Product Code. 15468809

Brand: Thermo Scientific Alfa Aesar L03071.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

A building block which is used in preparation of anthranilic acids possessing antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A building block which is used in preparation of anthranilic acids possessing antibacterial activity.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 68837-59-2
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00040905
InChI Key RVCJOGNLYVNRDN-UHFFFAOYSA-N
Synonym 2-methyl-4-bromobenzoic acid, 4-bromo-o-toluic acid, 4-bromo-2-methyl-benzoic acid, benzoic acid, 4-bromo-2-methyl, synquest 2721-9-x6, 4-bromo-2-methylbenz, 4-bromo-2-methylbenzoicacid, pubchem4029, 5-bromo-2-carboxytoluene, acmc-1baz5Show MoreShow Less
PubChem CID 99570
IUPAC Name 4-bromo-2-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)Br)C(=O)O

Spezifikation

Melting Point 182°C to 184°C
Quantity 100 g
Beilstein 2249816
Solubility Information Insoluble in water.
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-2-methylbenzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberI16T031 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL03071.06,L03071.03,L03071.14,L03071.22
Lot Number10148778 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL03071.06,L03071.03,L03071.14,L03071.22
Lot Number10125703 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL03071.06,L03071.03,L03071.14,L03071.22
Lot NumberL01Q074 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL03071.06,L03071.03,L03071.14,L03071.22
Lot NumberF13R024 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL03071.06,L03071.03,L03071.14,L03071.22

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
4-Bromo-2-methylbenzoic acid, 98+%, Thermo Scientific Chemicals >

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt