Learn More
4-(Bromomethyl)benzenesulfonyl chloride, 95%, Thermo Scientific Chemicals
354.00 SEK - 4085.00 SEK
Chemical Identifiers
CAS | 66176-39-4 |
---|---|
Molecular Formula | C7H6BrClO2S |
Molecular Weight (g/mol) | 269.537 |
MDL Number | MFCD00156129 |
InChI Key | QXTQWYZHHMQSQH-UHFFFAOYSA-N |
Synonym | 4-bromomethyl benzenesulfonyl chloride, 4-bromomethyl benzene-1-sulfonyl chloride, 4-bromomethyl benzenesulfonylchloride, alpha-bromo-p-toluenesulphonyl chloride, alpha-bromo-p-toluenesulfonyl chloride, 4-bromomethyl benzenesulphonyl chloride, 4-bromomethyl benzene-1-sulfonylchloride, buttpark 94\04-08, 4-bromomethyl phenyl chlorosulfone, 4-bromomethylene benzenesulfonyl chloride |
PubChem CID | 2734409 |
IUPAC Name | 4-(bromomethyl)benzenesulfonyl chloride |
SMILES | C1=CC(=CC=C1CBr)S(=O)(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15463017
|
Thermo Scientific Alfa Aesar
A10120.03 |
1 g | N/A | ||||||
11499362
|
Thermo Scientific Alfa Aesar
A10120.06 |
5 g |
1145.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409372
|
Thermo Scientific Alfa Aesar
A10120.14 |
25 g |
4085.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
66176-39-4 | |
269.537 | |
QXTQWYZHHMQSQH-UHFFFAOYSA-N | |
2734409 | |
C1=CC(=CC=C1CBr)S(=O)(=O)Cl |
C7H6BrClO2S | |
MFCD00156129 | |
4-bromomethyl benzenesulfonyl chloride, 4-bromomethyl benzene-1-sulfonyl chloride, 4-bromomethyl benzenesulfonylchloride, alpha-bromo-p-toluenesulphonyl chloride, alpha-bromo-p-toluenesulfonyl chloride, 4-bromomethyl benzenesulphonyl chloride, 4-bromomethyl benzene-1-sulfonylchloride, buttpark 94\04-08, 4-bromomethyl phenyl chlorosulfone, 4-bromomethylene benzenesulfonyl chloride | |
4-(bromomethyl)benzenesulfonyl chloride |
Specifications
66176-39-4 | |
C7H6BrClO2S | |
UN3261 | |
Moisture sensitive | |
QXTQWYZHHMQSQH-UHFFFAOYSA-N | |
4-(bromomethyl)benzenesulfonyl chloride | |
2734409 | |
95% |
72°C to 75°C | |
MFCD00156129 | |
2364328 | |
4-bromomethyl benzenesulfonyl chloride, 4-bromomethyl benzene-1-sulfonyl chloride, 4-bromomethyl benzenesulfonylchloride, alpha-bromo-p-toluenesulphonyl chloride, alpha-bromo-p-toluenesulfonyl chloride, 4-bromomethyl benzenesulphonyl chloride, 4-bromomethyl benzene-1-sulfonylchloride, buttpark 94\04-08, 4-bromomethyl phenyl chlorosulfone, 4-bromomethylene benzenesulfonyl chloride | |
C1=CC(=CC=C1CBr)S(=O)(=O)Cl | |
269.537 | |
269.55 | |
4- (Bromomethyl)benzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.