Learn More
4-Cyclohexylaniline, 97%, Thermo Scientific Chemicals
0.05 SEK - 820.00 SEK
Chemical Identifiers
CAS | 6373-50-8 |
---|---|
Molecular Formula | C16H12Cl2N4OS |
Molecular Weight (g/mol) | 379.26 |
MDL Number | MFCD00001454 |
InChI Key | YDTTUXLKBBQRHA-UHFFFAOYSA-N |
Synonym | benzenamine, 4-cyclohexyl, 4-cyclohexylbenzenamine, aniline, p-cyclohexyl, p-cyclohexylaniline, 4-cyclohexylphenyl amine, 4-cyclohexylphenylamine, 4-cyclohexylanilin, 4-cyclohexyl-aniline, acmc-20aivf, p-cyclohexylphenylamine |
PubChem CID | 80764 |
IUPAC Name | 4-cyclohexylaniline |
SMILES | CC1=CC=C(C=C1)C1=NN=C(NC(=O)NC2=CC=C(Cl)C=C2Cl)S1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11322056
|
Thermo Scientific Alfa Aesar
L03184.03 |
1 g |
820.00 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11332056
|
Thermo Scientific Alfa Aesar
L03184.06 |
5 g |
0.05 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Cyclohexylaniline is used as a starting material for the preparation of 1-cyclohexyl-4-nitroso-benzene. It is used as a reactant in nucleophilic acylation to form N-(4-cyclohexylphenyl)acetamide by reacting with acetyl halides. It is also involved in the synthesis of N-(4-cyclohexylphenyl)-2-morpholino-acetamide by reaction with 2-morpholinoacetyl fluoride. Further, it reacts with 4-bromo-4-oxo-butanoic acid to get 4-[(4-cyclohexylphenyl)amino]-4-oxo-butanoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Cyclohexylaniline is used as a starting material for the preparation of 1-cyclohexyl-4-nitroso-benzene. It is used as a reactant in nucleophilic acylation to form N-(4-cyclohexylphenyl)acetamide by reacting with acetyl halides. It is also involved in the synthesis of N-(4-cyclohexylphenyl)-2-morpholino-acetamide by reaction with 2-morpholinoacetyl fluoride. Further, it reacts with 4-bromo-4-oxo-butanoic acid to get 4-[(4-cyclohexylphenyl)amino]-4-oxo-butanoic acid.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
6373-50-8 | |
379.26 | |
YDTTUXLKBBQRHA-UHFFFAOYSA-N | |
80764 | |
CC1=CC=C(C=C1)C1=NN=C(NC(=O)NC2=CC=C(Cl)C=C2Cl)S1 |
C16H12Cl2N4OS | |
MFCD00001454 | |
benzenamine, 4-cyclohexyl, 4-cyclohexylbenzenamine, aniline, p-cyclohexyl, p-cyclohexylaniline, 4-cyclohexylphenyl amine, 4-cyclohexylphenylamine, 4-cyclohexylanilin, 4-cyclohexyl-aniline, acmc-20aivf, p-cyclohexylphenylamine | |
4-cyclohexylaniline |
Specifications
6373-50-8 | |
166°C (13 mmHg) | |
MFCD00001454 | |
benzenamine, 4-cyclohexyl, 4-cyclohexylbenzenamine, aniline, p-cyclohexyl, p-cyclohexylaniline, 4-cyclohexylphenyl amine, 4-cyclohexylphenylamine, 4-cyclohexylanilin, 4-cyclohexyl-aniline, acmc-20aivf, p-cyclohexylphenylamine | |
YDTTUXLKBBQRHA-UHFFFAOYSA-N | |
4-cyclohexylaniline | |
80764 | |
97% |
52°C to 56°C | |
C16H12Cl2N4OS | |
2965799 | |
Insoluble in water. | |
CC1=CC=C(C=C1)C1=NN=C(NC(=O)NC2=CC=C(Cl)C=C2Cl)S1 | |
379.26 | |
175.28 | |
4-Cyclohexylaniline |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'einecsNumber' : 228-918-9
missing translation for 'tsca' : No