missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-methylbenzaldehyde, 97%, Thermo Scientific Chemicals

576.30 SEK - 1940.74 SEK

Chemical Identifiers

CAS 52289-54-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD02261771
InChI Key WICYVKGMEJSDAO-UHFFFAOYSA-N
Synonym 2-methyl-4-methoxybenzaldehyde, 4-methoxy-2-methyl-benzaldehyde, benzaldehyde, 4-methoxy-2-methyl, 2-methyl-p-anisaldehyde, 2-methyl-anisaldehyde, acmc-1aoyo, 4-methoxy-2-methylbenzaldehyd, 4-methoxy-2-methylbenzaldehyde, 3-furane-2-amido benzoic acid
PubChem CID 283285
IUPAC Name 4-methoxy-2-methylbenzaldehyde
SMILES CC1=C(C=CC(=C1)OC)C=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10718815
View Documents
Thermo Scientific Acros
427020010
1 g Glass bottle
576.30 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
10526694
View Documents
Thermo Scientific Acros
427020050
5 g Glass bottle
1940.74 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

52289-54-0
150.18
WICYVKGMEJSDAO-UHFFFAOYSA-N
283285
CC1=C(C=CC(=C1)OC)C=O
C9H10O2
MFCD02261771
2-methyl-4-methoxybenzaldehyde, 4-methoxy-2-methyl-benzaldehyde, benzaldehyde, 4-methoxy-2-methyl, 2-methyl-p-anisaldehyde, 2-methyl-anisaldehyde, acmc-1aoyo, 4-methoxy-2-methylbenzaldehyd, 4-methoxy-2-methylbenzaldehyde, 3-furane-2-amido benzoic acid
4-methoxy-2-methylbenzaldehyde

Specifications

52289-54-0
100.0
1.1070g/mL
>110°C
96% min. (GC)
CH30C6H3(CH3)CHO
1.107
WICYVKGMEJSDAO-UHFFFAOYSA-N
4-methoxy-2-methylbenzaldehyde
283285
97%
4-Methoxy-2-methylbenzaldehyde
96.0
Colorless to Yellow
262°C to 267°C
Authentic
C9H10O2
MFCD02261771
2-methyl-4-methoxybenzaldehyde, 4-methoxy-2-methyl-benzaldehyde, benzaldehyde, 4-methoxy-2-methyl, 2-methyl-p-anisaldehyde, 2-methyl-anisaldehyde, acmc-1aoyo, 4-methoxy-2-methylbenzaldehyd, 4-methoxy-2-methylbenzaldehyde, 3-furane-2-amido benzoic acid
CC1=C(C=CC(=C1)OC)C=O
150.18
150.18
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

Product Identifier
  • 4-Methoxy-2-methylbenzaldehyde

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H371-May cause damage to organs.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only