missing translation for 'onlineSavingsMsg'
Learn More
4-n-Hexylaniline, 98%, Thermo Scientific Chemicals Product Code.: 11401150

4-n-Hexylaniline, 98%, Thermo Scientific Chemicals

Product Code. 11401150
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11401150

Brand: Thermo Scientific Alfa Aesar H26297.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Hexylaniline is used in the fabrication of RP/ion-exchange, mixed-mode, monolithic materials for capillary LC. The product is dye stuff intermediate, yet it may use to manufacture the medicine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hexylaniline is used in the fabrication of RP/ion-exchange, mixed-mode, monolithic materials for capillary LC. The product is dye stuff intermediate, yet it may use to manufacture the medicine

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33228-45-4
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.291
MDL Number MFCD00007927
InChI Key OVEMTTZEBOCJDV-UHFFFAOYSA-N
Synonym p-hexylaniline, 4-n-hexylaniline, benzenamine, 4-hexyl, aniline, p-hexyl, 4-hexylbenzenamine, p-n-hexylaniline, unii-u3c0879zgc, 4-hexylphenylamine, 4-n-hexylaniline;, acmc-209hzhShow MoreShow Less
PubChem CID 36382
IUPAC Name 4-hexylaniline
SMILES CCCCCCC1=CC=C(C=C1)N

Specifications

Density 0.919
Boiling Point 283°C to 285°C
Flash Point >110°C (230°F)
Refractive Index 1.525
Quantity 25 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 177.29
Percent Purity 98%
Chemical Name or Material 4-n-Hexylaniline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10228461 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26297.14,H26297.06
Lot Number10236618 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26297.14,H26297.06
Lot Number10171416 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26297.14,H26297.06
Lot Number10203442 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26297.14,H26297.06
Lot Number10196259 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26297.14,H26297.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-n-Hexylaniline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.