Learn More
4-n-Hexylaniline, 98%, Thermo Scientific Chemicals Product Code.: 11491140
4-n-Hexylaniline, 98%, Thermo Scientific Chemicals
Product Code. 11491140
5 g
Click to view available options
Quantity:
5 g
25 g
Product Code. 11491140
Brand: Thermo Scientific Alfa Aesar H26297.06
Request Bulk or Custom Format
4-Hexylaniline is used in the fabrication of RP/ion-exchange, mixed-mode, monolithic materials for capillary LC. The product is dye stuff intermediate, yet it may use to manufacture the medicine
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hexylaniline is used in the fabrication of RP/ion-exchange, mixed-mode, monolithic materials for capillary LC. The product is dye stuff intermediate, yet it may use to manufacture the medicine
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 33228-45-4
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.291
MDL Number MFCD00007927
InChI Key OVEMTTZEBOCJDV-UHFFFAOYSA-N
Synonym p-hexylaniline, 4-n-hexylaniline, benzenamine, 4-hexyl, aniline, p-hexyl, 4-hexylbenzenamine, p-n-hexylaniline, unii-u3c0879zgc, 4-hexylphenylamine, 4-n-hexylaniline;, acmc-209hzhShow MoreShow Less
PubChem CID 36382
IUPAC Name 4-hexylaniline
SMILES CCCCCCC1=CC=C(C=C1)N
Specifications
Density 0.919
Boiling Point 283°C to 285°C
Flash Point >110°C (230°F)
Refractive Index 1.525
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 177.29
Percent Purity 98%
Chemical Name or Material 4-n-Hexylaniline
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10236618 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26297.14,H26297.06
Lot Number10228461 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26297.14,H26297.06
Lot Number10171416 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26297.14,H26297.06
Lot Number10203442 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26297.14,H26297.06
Lot Number10196259 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26297.14,H26297.06
RUO – Research Use Only
Product Content Correction
Product Title
4-n-Hexylaniline, 98%, Thermo Scientific Chemicals >