Learn More
Brady™ 4500 Series Thermal Transfer Ribbons
Environmental resistance against smearing and chemicals. Brady 4500 Series Thermal Transfer Ribbons have wax/resin formulation and available in variuos sizes and colors.
Brand:  Brady™ 055733
Additional Details : Weight : 0.28210kg
 View more versions of this product
Product Code. 11518464
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Offers environmental resistance against smearing and chemicals
  • Sharp-edge print quality to maximize bar code readability
Specifications
Specifications
Printer Ribbon
Brady 2461, Thermal Transfer Printer, Brady 6441, BBP81 Label Printer, BradyPrinter PR Plus Series Label Printer, Brady 300X-Plus II, Brady 360X-Plus II, Brady 3481, Tagus T200, Brady 300MVP Plus, Brady 200MVP Plus, Brady 300X-Plus II, Brady 360X-Plus II, Brady 600X-Plus II, Tagus T200, Tagus T300
300.00 m
25.4 mm I.D.
Blue 4500 Series Thermal Transfer Printer Ribbon
1 Ea.
Blue
Wax
83 mm
299 m x 83.06 mm (984 ft. x 3.27 in.)
Ink side out