Learn More
Hamilton™ 700 Series Digital Cemented Needle Custom Syringes
This series of syringes is ideal for dispensing volumes from 0.5 up to 500μL.
Brand:  Hamilton™ DS80608
Product Code. 11528054
  • 19795.00 SEK / Each
Explore more special offers
Description
Description
  • The original hand-fitted Hamilton™ syringe
  • Stainless steel plunger is manufactured to fit the glass barrel with a tolerance less than three thousandths of a millimeter
  • Unsurpassed syringe life
Specifications
Specifications
Not Autoclavable
Custom
7.75 mm
Microliter Syringe
50°C
10°C
100 μL
Cemented Needle Special
Borosilicate Glass, Stainless Steel
1.46 mm
Ideal for dispensing volumes from 0.5 μL up to 500 μL
2000 psig (137.9 bar)
710 SN
1/Ea.
Custom