Learn More
A-867744, MedChemExpress
MedChemExpress A-867744 is a highly potent and selective type II positive allosteric modulator (PAM) of the alpha7 nicotinic acetylcholine receptors (nAChR) with an EC50 of 1.0 μM.
Brand:  Medchem Express HY-12149
Encompass_Preferred
 View more versions of this product
Product Code. 16444998
  • 2007.32 SEK / 10mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
A-867744 is a highly potent and selective type II positive allosteric modulator (PAM) of the alpha7 nicotinic acetylcholine receptors (nAChR) with an EC50 of 1.0 μM.
Specifications
Specifications
A-867744
Off-White
C20H19ClN2O3S
DMSO : ≥ 100 mg/mL (248.21 mM)
402.89
95.06%
Solid
1000279-69-5
Research
10 mg
O=S(C1=CC=C(C=C1)N2C(C)=C(C=C2C3=CC=C(C=C3)Cl)C(CC)=O)(N)=O
402.89
Research
Neuroscience-Neuromodulation
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
A-867744, MedChemExpress