Learn More
Avadomide, MedChemExpress
MedChemExpress Avadomide (CC 122) is an orally active cereblon modulator. Avadomide modulates cereblon E3 ligase activity and induces apoptosis of diffuse large B-cell lymphoma (DLBCL) cell lines. Avadomide exhibits potent antitumor and immunomodulatory activities.
Brand: Medchem Express HY-100507
Description
Avadomide (CC 122) is an orally active cereblon modulator. Avadomide modulates cereblon E3 ligase activity and induces apoptosis of diffuse large B-cell lymphoma (DLBCL) cell lines. Avadomide exhibits potent antitumor and immunomodulatory activities.Specifications
Avadomide | |
Gray | |
C14H14N4O3 | |
CC 122 | |
O=C(C(N1C(C)=NC2=C(C(N)=CC=C2)C1=O)CC3)NC3=O | |
286.29 | |
Research | |
Cancer-programmed cell death |
1015474-32-4 | |
Research | |
100 mg | |
DMSO : 33.33 mg/mL (116.42 mM; Need ultrasonic) | |
286.29 | |
99.5% | |
Solid |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only