missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Adapters with NS Stopcock, Bent

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

426.00 SEK - 473.00 SEK

Specifications

Includes Bent glass olive (90°)
Material Borosilicate Glass (3.3)
View More Specs

Products 3
Product Code Brand Standard Taper Joint No. Price Quantity & Availability  
Product Code Brand Standard Taper Joint No. Price Quantity & Availability  
10264481
View Documents Promotion Details
Lenz Laborglasintrumente
05105004
NS 29/32
473.00 SEK
Each
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
 
10090481
View Documents Promotion Details
Lenz Laborglasintrumente
05105002
NS 19/26
426.00 SEK
Each
Estimated Shipment: 10-12-2024
to see stock.
Add to basket
 
10396021
View Documents Promotion Details
Lenz Laborglasintrumente
05105003
NS 24/29
473.00 SEK
Each
Estimated Shipment: 10-12-2024
to see stock.
Add to basket
 
Description

Description

  • Adapters with NS socket
  • Bent glass olive (90°) and NS stopcock with solid glass plug
  • Bore: 2,5 mm
  • Made of DURAN™ tubing
Specifications

Specifications

Bent glass olive (90°)
Borosilicate Glass (3.3)
Videos
SDS
Documents

Documents

Product Certifications