Learn More
BTX™ AgilePulse™ In Vivo Adapter Box
AgilePulse™ In Vivo Adapter Box for use with AgilePulse™ In Vivo system.
Brand: BTX™ 470091
Additional Details : Weight : 0.96000kg
Description
- For connection to electroporation generators
Specifications
In-Vivo Drug or Gene Delivery, Gene Therapy, Protein Delivery | |
BTX Electroporation | |
AgilePulse In Vivo Adapter Box, for generators with SN AP-4069-ID / AP-5031-IM and after | |
Adapter Box Only | |
Electrode Connector Cable | |
Square Wave |
AgilePulse Generators with SN AP-4069-ID/AP-5031-IM and After | |
Cables & Adapters | |
Touchscreen | |
Non-sterile | |
Touchscreen |
Intended for Research use only. Not for use in diagnostic or clinical procedures.