missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ AgilePulse™ In Vivo Adapter Box
AgilePulse™ In Vivo Adapter Box for use with AgilePulse™ In Vivo system.
1065.00 SEK - 1430.00 SEK
Specifications
For Use With (Application) | In-Vivo Drug or Gene Delivery, Gene Therapy, Protein Delivery |
---|---|
Product Line | BTX Electroporation |
Includes | Adapter Box Only |
Sterility | Non-sterile |
Pulse Form | Square Wave |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15457380
|
BTX™
470090 |
AgilePulse Generators with SN AP-4068-ID/AP-5030-IM and Before |
1065.00 SEK
Each |
Estimated Shipment: 18-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16243931
|
BTX™
470091 |
AgilePulse Generators with SN AP-4069-ID/AP-5031-IM and After |
1430.00 SEK
Each |
Estimated Shipment: 18-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For connection to electroporation generators
Specifications
In-Vivo Drug or Gene Delivery, Gene Therapy, Protein Delivery | |
Adapter Box Only | |
Square Wave |
BTX Electroporation | |
Non-sterile |
Intended for Research use only. Not for use in diagnostic or clinical procedures.