missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5122258
This item is not returnable.
View return policy
Description
AIM1 Polyclonal antibody specifically detects AIM1 in Human samples. It is validated for Western Blot.Specifications
AIM1 | |
Polyclonal | |
Unconjugated | |
CRYBG1 | |
absent in melanoma 1; absent in melanoma 1 protein; AIM1; beta/gamma crystallin domain-containing protein 1; beta-gamma crystallin domain containing 1; CRYBG1; ST4; suppression of tumorigenicity 4 (malignant melanoma) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
202 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9Y4K1 | |
CRYBG1 | |
Pro1502-Met1719 (Accession Q9Y4K1), with N-terminal His Tag | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |