Learn More
Thermo Scientific™ Aluminum Crimp Seals with Prefitted Septa for 8mm Crimp Top Vials
Thermo Scientific™ National Aluminum Crimp Seals provide a secure, leak-resistant seal that you can count on.
Brand: Thermo Scientific™ C4008-10
This item is not returnable.
View return policy
Description
Thermo Scientific™ National Aluminum Crimp Seals provide a secure, leak-resistant seal that you can count on.
- Available in a variety of colors and septum types
- Aluminum seals must be applied with a crimping tool
Specifications
Integral Polyethylene septa; Clear polyethylene; Snap-it cap | |
Polyethylene | |
8 mm |
Clear Septa, Clear | |
Clear Integral Polyethylene septa | |
8 mm Crimp Top Vials |