Learn More
Thermo Scientific™ Aluminum Crimp Seals with Prefitted Septa for 8mm Crimp Top Vials
Thermo Scientific™ National Aluminum Crimp Seals provide a secure, leak-resistant seal that you can count on.
92.90 SEK - 880.00 SEK
Specifications
Closure Size 8 mm
For Use With (Equipment) 8 mm Crimp Top Vials
View More Specs
Products 3
Product Code Brand Color Septum Price Quantity & Availability  
Product Code Brand Color Septum Price Quantity & Availability  
12433588
View Documents
Thermo Scientific™
C4008-10
Clear Septa, Clear Clear Integral Polyethylene septa This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10087124
View Documents
Thermo Scientific™
C4008-6A
White, Silver White Virgin PTFE septa This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12493598
View Documents
Thermo Scientific™
C4008-98B
Red, Blue Clear PTFE/red rubber This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Thermo Scientific™ National Aluminum Crimp Seals provide a secure, leak-resistant seal that you can count on.
  • Available in a variety of colors and septum types
  • Aluminum seals must be applied with a crimping tool
Specifications
Specifications
8 mm
8 mm Crimp Top Vials