Learn More
Thermo Scientific™ Aluminum Crimp Seals with Prefitted Septa for 8mm Crimp Top Vials
Thermo Scientific™ National Aluminum Crimp Seals provide a secure, leak-resistant seal that you can count on.
Brand:  Thermo Scientific™ C4008-98B
 View more versions of this product
Product Code. 12493598
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Thermo Scientific™ National Aluminum Crimp Seals provide a secure, leak-resistant seal that you can count on.
  • Available in a variety of colors and septum types
  • Aluminum seals must be applied with a crimping tool
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear PTFE/red rubber septa; Blue
PTFE, Rubber
8 mm
Red, Blue
Clear PTFE/red rubber
8 mm Crimp Top Vials