missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Thiocyanate, 99.99% Trace Metals Basis, Honeywell™

99.99% trace metals basis

1373.64 SEK - 4640.56 SEK

Chemical Identifiers

CAS 1762-95-4
Molecular Formula CH4N2S
Molecular Weight (g/mol) 76.117
MDL Number MFCD00011428
InChI Key SOIFLUNRINLCBN-UHFFFAOYSA-N
Synonym ammonium thiocyanate, ammoniumrhodanid, thiocyanic acid, ammonium salt, ammonium isothiocyanate, ammonium rhodanate, weedazol tl, trans-aid, ammonium sulfocyanate, ammonium sulfocyanide, rhodanine, ammonium saltShow MoreShow Less
PubChem CID 15666
IUPAC Name azanium;thiocyanate
SMILES C(#N)[S-].[NH4+]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15655510
SDS View Documents
Honeywell Research Chemicals
431354-50G
50 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15645510
SDS View Documents
Honeywell Research Chemicals
431354-250G
250 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P273-P280

missing translation for 'rtecsNumber' : 217-175-6

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Thiocyanate, 99.99% Trace Metals Basis, Honeywell™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.