Learn More
STEMCELL Technologies Anacardic Acid
Epigenetic modifier; Inhibits histone acetyltransferase (HAT)
Brand: STEMCELL Technologies 73194
This item is not returnable.
View return policy
Description
- Anacardic Acid is a 6-alkyl salicyclic acid that inhibits the histone acetyltransferase (HAT) activity of the transcription co-activators p300 and p300/CREB-binding protein-associated factor (pCAF; IC50 values of 8.5 and 5 μM, respectively; Balasubramanyam et al.) Anacardic Acid inhibits HAT-dependent transcription and protein SUMOylation (Cui et al.; Fukuda et al.) In addition, Anacardic Acid is an activator of Aurora kinase A-mediated phosphorylation of Histone H3 (Kishore et al.) At higher concentrations (25 μM), Anacardic Acid suppresses NF-κB activation and inhibits IκB-α phosphorylation (Sung et al.)
Specifications
16611-84-0 | |
5 mg | |
C22H36O3 | |
≥98% |