missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Anacardic Acid

Epigenetic modifier; Inhibits histone acetyltransferase (HAT)

Specifications

CAS 16611-84-0
Molecular Formula C22H36O3
Purity ≥98%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17170182
Encompass_Preferred
View Documents Promotion Details
STEMCELL Technologies
73192
1 mg N/A
View alternative products
Save Up to 
»
N/A
17180182
Encompass_Preferred
View Documents Promotion Details
STEMCELL Technologies
73194
5 mg N/A
View alternative products
Save Up to 
»
N/A
Description

Description

  • Anacardic Acid is a 6-alkyl salicyclic acid that inhibits the histone acetyltransferase (HAT) activity of the transcription co-activators p300 and p300/CREB-binding protein-associated factor (pCAF; IC50 values of 8.5 and 5 μM, respectively; Balasubramanyam et al.) Anacardic Acid inhibits HAT-dependent transcription and protein SUMOylation (Cui et al.; Fukuda et al.) In addition, Anacardic Acid is an activator of Aurora kinase A-mediated phosphorylation of Histone H3 (Kishore et al.) At higher concentrations (25 μM), Anacardic Acid suppresses NF-κB activation and inhibits IκB-α phosphorylation (Sung et al.)
Specifications

Specifications

16611-84-0
C22H36O3
≥98%
Videos
SDS
Documents

Documents

Product Certifications