missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Annexin V, APC
A cellular protein for detection of apoptotic cells, PE-conjugated
2473.20 SEK - 5728.05 SEK
Specifications
For Use With (Application) | Flow Cytometry |
---|---|
Conjugate | APC (Allophycocyanin) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17107561
|
STEMCELL Technologies
100-0328 |
25 Tests |
2473.20 SEK
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17117561
|
STEMCELL Technologies
100-0329 |
100 Tests |
5728.05 SEK
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Annexin V is a member of the annexin family of proteins that bind to membrane phospholipids in the presence of calcium
- This dye has high affinity for phosphatidylserine (PS) that is present in the inner leaflet of the plasma membrane
- During early-stage cell apoptosis, PS is translocated from the inner to the outer leaflet of the plasma membrane, exposing it to the external environment
- Annexin V, a characteristic marker for early cell apoptosis, detects the translocation of PS to the external environment
- Annexin V is used along with viability dyes such as 7-AAD (Catalog #75001) or Propidium Iodide (Catalog #75002)
- The process of PS translocation occurs prior to the loss of membrane integrity
- Therefore, as cells progress through apoptosis and towards necrosis, the cell membrane is compromised and consequently, viability dyes pass into the cell
- Thus, cells undergoing early apoptosis stain positive for Annexin V and negative for viability dyes, while apoptotic death or necrosis is characterized by positive staining for both Annexin V and the viability dye
Specifications
Flow Cytometry | |
APC (Allophycocyanin) |