missing translation for 'onlineSavingsMsg'
Learn More

MEK1 Mouse, Unlabeled, Clone: 25/MEK1, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610122

 View more versions of this product

Product Code. 15895269

  • 4105.00 SEK / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

MEK1 (MapK/ERK Kinase 1) is a 45kDa member of the MEK family of dual specificity kinases. MEK is activated by a variety of cellular serine/threonine kinases including c-Raf, A-Raf, c-mos, and MEK Kinase-1. Activated MEK phosphorylates MAP kinase (ERK) at threonine and tyrosine residues. This results in activation of ERK and its signaling pathway. MEK is highly specific for ERK and various MEKs preferentially phosphorylate individual ERK isoforms. MEK1 only activates ERK1 and ERK2. This specificity may result from variations in
ERK regions that are known as the phosphorylation lip and kinase backbone. MEK's localization is cytoplasmic, but mitogenic stimulation induces a mass translocation to the nucleus. Mechanisms behind this nuclear translocation remain unknown. However, MEK contains an N-terminal nuclear export signal (NES) that mediates its rapid exodus from the nucleus and restores its unstimulated cellular distribution.

Host Species: Mouse
Clone: 25
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Human MEK1 aa. 2-124
Formula Weight [Chemical]: 45kDa

Bioimaging, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

MEK1
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human MEK1 aa. 2-124
150 μg
Cell Biology
Canine, Chicken, Frog, Human, Mouse, Rat
IgG2a
Western Blot
25/MEK1
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.