Learn More
Artefenomel, MedChemExpress
MedChemExpress Artefenomel (OZ439) is a synthetic antimalarial agent with the artemisinin pharmacophore. Artefenomel (OZ439) is a long-acting artemisinin-related agent.
Brand:  Medchem Express HY-16762
Encompass_Preferred
 View more versions of this product
Product Code. 16437176
  • 53855.25 SEK / 100mg
Explore more special offers
Description
Description
Artefenomel (OZ439) is a synthetic antimalarial agent with the artemisinin pharmacophore. Artefenomel (OZ439) is a long-acting artemisinin-related agent.
Specifications
Specifications
Artefenomel
1029939-86-3
100 mg
C28H39NO5
OZ439
[H][C@@]1(C[C@H](C2)C3)C4([C@@]3([H])C[C@H]2C1)O[C@]5(OO4)CC[C@H](CC5)C6=CC=C(C=C6)OCCN7CCOCC7
469.61
Research
Solid
Off-White
Research
COVID-19-immunoregulation
DMSO : 4.2 mg/mL (8.94 mM; Need ultrasonic)
469.61
98.16%
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Artefenomel, MedChemExpress